missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific Chemicals

Catalog Number 11915961
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
213343-65-8
C22H40BF4P2Rh
556.22
MFCD09038435
CVUPEPGZCZHSOF-BDNLEJQTNA-N
1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, s,s-me-bpe-rh, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene-rhodium i tetrafluoroborate
12964417
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C[C@H]1CC[C@H](C)P1CCP1[C@@H](C)CC[C@@H]1C
This item is not returnable. View return policy
213343-65-8
C22H40BF4P2Rh
556.22
MFCD09038435
CVUPEPGZCZHSOF-BDNLEJQTNA-N
1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, s,s-me-bpe-rh, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene-rhodium i tetrafluoroborate
12964417
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C[C@H]1CC[C@H](C)P1CCP1[C@@H](C)CC[C@@H]1C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 152.0°C to 156.0°C
Color Brown to Orange
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 100 mg
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;(2S,5S)-1-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethylphospholane;rhodium;tetrafluoroborate
Formula Weight 556.21
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis((2S,5S)-2,5-Dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'