Learn More
1,2-Bis((2R,5R)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, ACROS Organics™
Brand: Acros Organics 435221000
Code : 29
Additional Details : CAS Number : 305818-67-1 Weight : 0.05010kg
Quantity | 100mg |
---|
Chemical Identifiers
1,2-Bis((2R,5R)-2,5-Dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate | |
96.0 | |
97% | |
556.21 |
305818-67-1 | |
100.0 | |
BF4·HRh | |
MFCD09265087 |
Specifications
1,2-Bis((2R,5R)-2,5-Dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate | |
96.0 | |
45.6 to 49.4% C | |
(1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure, NMR (19F-NMR), Consistent with structure | |
97% | |
BF4·HRh | |
MFCD09265087 | |
100mg |
305818-67-1 | |
100.0 | |
Authentic | |
Powder | |
97% | |
556.21 | |
Orange to red to brown |