missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific Chemicals

Catalog Number 10812261
Click to view available options
Quantity:
100 mg
250 mg
This item is not returnable. View return policy
228121-39-9
C30H48BF4P2Rh
660.37
MFCD01862464
DSYBEQKPSKLNMC-RTXTXWNLNA-N
11549211
cyclooctane;(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;tetrafluoroborate
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC[C@@H]1CC[C@@H](CC)P1C1=C(C=CC=C1)P1[C@H](CC)CC[C@H]1CC
This item is not returnable. View return policy
228121-39-9
C30H48BF4P2Rh
660.37
MFCD01862464
DSYBEQKPSKLNMC-RTXTXWNLNA-N
11549211
cyclooctane;(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;tetrafluoroborate
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC[C@@H]1CC[C@@H](CC)P1C1=C(C=CC=C1)P1[C@H](CC)CC[C@H]1CC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Brown to Orange
Elemental Analysis 52.4 to 56.8% C
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 100 mg
Formula Weight 660.37
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,2-Bis((2R,5R)-2,5-Diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'