missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzenedithiol, 97%, Thermo Scientific Chemicals

Catalog Number p-7045585
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
17534-15-5
C6H6S2
142.23
MFCD00004835
JRNVQLOKVMWBFR-UHFFFAOYSA-N
1,2-benzenedithiol, benzenedithiol, 1,2-dimercaptobenzene, dithiocatechol, dimercaptobenzene, o-benzenedithiol, acmc-209e9y, 1,2-benzenedithiol 9ci, benzene-1,2-dithiol
69370
benzene-1,2-dithiol
SC1=C(S)C=CC=C1
This item is not returnable. View return policy
17534-15-5
C6H6S2
142.23
MFCD00004835
JRNVQLOKVMWBFR-UHFFFAOYSA-N
1,2-benzenedithiol, benzenedithiol, 1,2-dimercaptobenzene, dithiocatechol, dimercaptobenzene, o-benzenedithiol, acmc-209e9y, 1,2-benzenedithiol 9ci, benzene-1,2-dithiol
69370
benzene-1,2-dithiol
SC1=C(S)C=CC=C1

1,2-Benzenedithiol is used in preparation of schiff base, which is employed to make new series of copper(II) and zinc(II) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Benzenedithiol is used in preparation of schiff base, which is employed to make new series of copper(II) and zinc(II) complexes.

Solubility
Soluble in water, benzene, ethanol, diethyl ether, ethyl acetate.

Notes
Incompatible with bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 21°C to 25°C
Density 1.236
Boiling Point 239°C
Flash Point 103°C (217°F)
Odor Unpleasant
Refractive Index 1.658
Quantity 250 mg
UN Number UN3335
Beilstein 636154
Sensitivity Air sensitive
Solubility Information Soluble in water,benzene,ethanol,diethyl ether,ethyl acetate.
Formula Weight 142.24
Percent Purity 97%
Chemical Name or Material 1,2-Benzenedithiol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'