missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride, 98%

Brand:  Alfa Aesar™ H64862.14

Code : NEW

Additional Details : CAS Number : 130198-05-9 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15449206

  • £29.00 / 25g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride
285.81
4215444
130198-05-9
MFCD06658143

Specifications

1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride
Crystalline Powder
174° to 178°C (lit.)
MFCD06658143
25g
130198-05-9
98%
285.81
White
4215444
Videos
SDS