missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,6-Dichlorobenzyl)piperazine, 97%

Brand:  Alfa Aesar™ H55486.14

Code : NEW

Additional Details : CAS Number : 102292-50-2 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15441758

  • £557.14 / 25g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,6-Dichlorobenzyl)piperazine
102292-50-2
245.15
MFCD02736281

Specifications

1-(2,6-Dichlorobenzyl)piperazine
54° to 62°C
102292-50-2
97%
MFCD02736281
White to pale brown
25g
Solid
245.15
Videos
SDS