missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,6-Dichlorobenzyl)piperazine, 97%

Brand:  Alfa Aesar™ H55486.06

Code : NEW

Additional Details : CAS Number : 102292-50-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15431758

  • £124.02 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,6-Dichlorobenzyl)piperazine
102292-50-2
245.15
MFCD02736281

Specifications

1-(2,6-Dichlorobenzyl)piperazine
54° to 62°C
102292-50-2
97%
MFCD02736281
White to pale brown
5g
Solid
245.15
Videos
SDS