missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,5-Difluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61835.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15476806

  • £95.61 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,5-Difluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
316.23
MFCD26143300

Specifications

1-(2,5-Difluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
1g
97%
MFCD26143300
White
Solid
316.23
Videos
SDS