missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,5-Difluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64558.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15478976

  • £91.90 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,5-Difluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
332.23
MFCD18185765

Specifications

1-(2,5-Difluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
332.23
1g
97%
MFCD18185765
Videos
SDS