missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,5-Dichlorophenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62278.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15428828

  • £491.84 / 5g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,5-Dichlorophenyl)-3-(4-fluorophenyl)urea
299.13
MFCD00086050

Specifications

1-(2,5-Dichlorophenyl)-3-(4-fluorophenyl)urea
Solid
299.13
5g
97%
MFCD00086050
Videos
SDS