missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[2,5-Bis(trifluoromethyl)phenyl]-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61618.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15496646

  • £73.56 / 1g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

1-[2,5-Bis(trifluoromethyl)phenyl]-3-[4-(trifluoromethyl)phenyl]urea
416.24
MFCD26143293

Specifications

1-[2,5-Bis(trifluoromethyl)phenyl]-3-[4-(trifluoromethyl)phenyl]urea
97%
MFCD26143293
Solid
416.24
1g
Videos
SDS