missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1-[2,5-Bis(trifluoromethyl)phenyl]-3-(4-fluorophenyl)urea, 97%
Brand: Alfa Aesar™ H62610.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1-[2,5-Bis(trifluoromethyl)phenyl]-3-(4-fluorophenyl)urea | |
366.23 | |
MFCD26960678 |
Specifications
1-[2,5-Bis(trifluoromethyl)phenyl]-3-(4-fluorophenyl)urea | |
97% | |
MFCD26960678 |
Solid | |
366.23 | |
1g |