missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,5,6-Diepoxycyclooctane, 96%, Thermo Scientific™
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Specifications
CAS | 27035-39-8 |
CAS Min % | 95.0 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Boiling Point | 90.0°C (5.0 mmHg) |
Flash Point | 105°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C8H12O2 |
Show More |
Safety and Handling
Product Identifier
- 1,2,5,6-Diepoxycyclooctane
- Warning
- Carcinogenicity Category 2
- Germ cell mutagenicity Category 2
- H351-Suspected of causing cancer.
- H341-Suspected of causing genetic defects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'