missing translation for 'onlineSavingsMsg'
Learn More

1,2:5,6-Di-O-isopropylidene-D-mannitol, 97%, ACROS Organics™

Brand:  Acros Organics 393330010

Code : 29

Additional Details : CAS Number : 1707-77-3 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10637970

  • £11.40 / 1g


Estimated Shipment
22-06-2018
Add to basket

Specifications

Chemical Identifiers

1, 2:5, 6-Di-O-isopropylidene-D-mannitol
96.0
97%
C12H22O6
D-Mannitol diacetonide
1707-77-3
100.0
97%
262.3

Specifications

1, 2:5, 6-Di-O-isopropylidene-D-mannitol
96.0
Authentic
+5° to +7° (20°C, 589nm) c=5, chloroform
Crystalline Powder
White
C12H22O6
262.3
1707-77-3
100.0
Glass bottle
105.0° to 114.0°C
97%
97%
1g
D-Mannitol diacetonide
Videos
SDS