missing translation for 'onlineSavingsMsg'
Learn More

1,2:5,6-Di-O-isopropylidene-D-mannitol, 97%, Thermo Scientific Chemicals

Catalog Number 10647970
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1707-77-3
C12H22O6
262.3
ODYBCPSCYHAGHA-ZYUZMQFOSA-N
1,2:5,6-di-o-isopropylidene-d-mannitol, d-mannitol diacetonide, 1,2:5,6-bis-o-1-methylethylidene-d-mannitol, d-mannitol 1,2:5,6-bis-acetonide, 1s,2s-1,2-bis 4r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1s,2s-1,2-bis r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1,2,5,6-di-o-isopropylidene-d-mannitol, mannitol diacetonide, diacetone-d-mannitol, zlchem 213
96011
(1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol
CC1(OCC(O1)C(C(C2COC(O2)(C)C)O)O)C
This item is not returnable. View return policy
1707-77-3
C12H22O6
262.3
ODYBCPSCYHAGHA-ZYUZMQFOSA-N
1,2:5,6-di-o-isopropylidene-d-mannitol, d-mannitol diacetonide, 1,2:5,6-bis-o-1-methylethylidene-d-mannitol, d-mannitol 1,2:5,6-bis-acetonide, 1s,2s-1,2-bis 4r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1s,2s-1,2-bis r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1,2,5,6-di-o-isopropylidene-d-mannitol, mannitol diacetonide, diacetone-d-mannitol, zlchem 213
96011
(1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol
CC1(OCC(O1)C(C(C2COC(O2)(C)C)O)O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 105.0°C to 114.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min
Packaging Glass bottle
Quantity 5 g
Specific Rotation +5° to +7° (20°C, 589nm) c=5, chloroform
Formula Weight 262.3
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1, 2:5, 6-Di-O-isopropylidene-D-mannitol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'