missing translation for 'onlineSavingsMsg'
Learn More

1,2:5,6-Di-O-isopropylidene-alpha-D-allo-furanose, 98%, Thermo Scientific Chemicals

Catalog Number p-3757719
Click to view available options
Quantity:
500 mg
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2595-05-3
C12H20O6
260.28
MFCD00135634
KEJGAYKWRDILTF-VVULQXIFSA-N
1,2:5,6-di-o-isopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyltetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 1,2,5,6-di-o-isopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-4r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 1,2:5,6-bis-o-1-methylethylidene, 5,6-di-o-isopropylidene-alpha-d-allofuranose, 1,2:5,6-di-o-isopropylidene-?-d-allofuranose, 1,2:5,6-di-o-isopropylidene-alpha-d-ribo-hexofuranose, 1-o,2-o:5-o,6-o-diisopropylidene-alpha-d-allofuranose
7157054
(3aR,5S,6R,6aR)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C
This item is not returnable. View return policy
2595-05-3
C12H20O6
260.28
MFCD00135634
KEJGAYKWRDILTF-VVULQXIFSA-N
1,2:5,6-di-o-isopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyltetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 1,2,5,6-di-o-isopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-4r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 1,2:5,6-bis-o-1-methylethylidene, 5,6-di-o-isopropylidene-alpha-d-allofuranose, 1,2:5,6-di-o-isopropylidene-?-d-allofuranose, 1,2:5,6-di-o-isopropylidene-alpha-d-ribo-hexofuranose, 1-o,2-o:5-o,6-o-diisopropylidene-alpha-d-allofuranose
7157054
(3aR,5S,6R,6aR)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 73°C to 77°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 500 mg
Specific Rotation Condition +37.50 (20.00°C c=1,chloroform)
Specific Rotation + 37.5
Formula Weight 260.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 2:5, 6-Di-O-isopropylidene-α-D-allo-furanose
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'