missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trimethoxybenzene, 98%, Thermo Scientific Chemicals

Catalog Number p-7080790
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
135-77-3
C9H12O3
168.192
MFCD00008360
AGIQIOSHSMJYJP-UHFFFAOYSA-N
benzene, 1,2,4-trimethoxy, unii-3ku3wd07ss, epa pesticide chemical code 040515, 3ku3wd07ss, hydroxyhydroquinone trimethyl ether, 2,4,5-trimethoxybenzene, acmc-1br3s, 1,3,4-trimethoxy benzene, dsstox_cid_15163, dsstox_rid_79243
67284
1,2,4-trimethoxybenzene
COC1=CC(=C(C=C1)OC)OC
This item is not returnable. View return policy
135-77-3
C9H12O3
168.192
MFCD00008360
AGIQIOSHSMJYJP-UHFFFAOYSA-N
benzene, 1,2,4-trimethoxy, unii-3ku3wd07ss, epa pesticide chemical code 040515, 3ku3wd07ss, hydroxyhydroquinone trimethyl ether, 2,4,5-trimethoxybenzene, acmc-1br3s, 1,3,4-trimethoxy benzene, dsstox_cid_15163, dsstox_rid_79243
67284
1,2,4-trimethoxybenzene
COC1=CC(=C(C=C1)OC)OC

1,2,4-Trimethoxybenzene is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4-Trimethoxybenzene is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store away from incompatible materials such as oxidizing agents, acids and bases. Use only in a chemical fumehood.
TRUSTED_SUSTAINABILITY
Density 1.128
Boiling Point 247°C
Flash Point >110°C (230°F)
Refractive Index 1.533
Quantity 10 g
Beilstein 2047579
Merck Index 14,1073
Solubility Information Not miscible in water.
Formula Weight 168.19
Percent Purity 98%
Chemical Name or Material 1,2,4-Trimethoxybenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'