missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,4-Dimethylphenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62393.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458908

  • £50.35 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,4-Dimethylphenyl)-3-(4-fluorophenyl)urea
258.29
MFCD00438028

Specifications

1-(2,4-Dimethylphenyl)-3-(4-fluorophenyl)urea
Solid
258.29
1g
97%
MFCD00438028
Videos
SDS