missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,4-Difluorophenyl)-3-phenylurea, 95%

Brand:  Alfa Aesar™ H61207.03

Code : NEW

Additional Details : CAS Number : 368-01-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15417718

  • £79.71 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,4-Difluorophenyl)-3-phenylurea
248.23
3357451
368-01-4
MFCD03386993

Specifications

1-(2,4-Difluorophenyl)-3-phenylurea
1g
Solid
248.23
3357451
White
368-01-4
95%
MFCD03386993
Videos
SDS