Alfa Aesar™ 1-(2,4-Difluorophenyl)-3-(4-fluorophenyl)urea, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H62424.03

1GR 1-(2,4-Difluorophenyl)-3-(4-fluorophenyl)urea, 97% 1g

UNSPSC: 12352116

Code: NEW

Additional Details:
Additional Details: Weight: 0.00100kg


Quantity 1g

Product Code. 15428928

Quantity Price
1 £ 74.73 / 1g
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1-(2,4-Difluorophenyl)-3-(4-fluorophenyl)urea
Formula Weight 266.22
MDL Number MFCD07092265

Specification

Formula Weight 266.22
Chemical Name or Material 1-(2,4-Difluorophenyl)-3-(4-fluorophenyl)urea
Physical Form Solid
Assay 97%
MDL Number MFCD07092265
Quantity 1g