missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,4-Difluorophenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62424.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15438928

  • £373.12 / 5g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,4-Difluorophenyl)-3-(4-fluorophenyl)urea
266.22
MFCD07092265

Specifications

1-(2,4-Difluorophenyl)-3-(4-fluorophenyl)urea
Solid
266.22
5g
97%
MFCD07092265
Videos
SDS