missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,4-Dichlorobenzyl)piperazine, 97%

Brand:  Alfa Aesar™ H55440.03

Code : NEW

Additional Details : CAS Number : 51619-56-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15461678

  • £31.60 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,4-Dichlorobenzyl)piperazine
51619-56-8
245.15
MFCD03407487

Specifications

1-(2,4-Dichlorobenzyl)piperazine
1g
1.255g/mL
Liquid
245.15
Pale yellow
51619-56-8
1.5710
97%
MFCD03407487
Videos
SDS