missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,4,6-Trimethylbenzyl)piperazine, 97%

Brand:  Alfa Aesar™ H55734.06

Code : NEW

Additional Details : CAS Number : 41717-26-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15422068

  • £103.80 / 5g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,4,6-Trimethylbenzyl)piperazine
41717-26-4
218.34
MFCD03094587

Specifications

1-(2,4,6-Trimethylbenzyl)piperazine
88° to 98°C
41717-26-4
97%
MFCD03094587
White to pale yellow
5g
Powder
218.34
Videos
SDS