missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrachlorobenzene, 98%, ACROS Organics™

Brand:  Acros Organics 157782500

Code : 29

Additional Details : CAS Number : 95-94-3 Weight : 0.30000kg


Packaging Glass bottle
Quantity 250g

 View more versions of this product

Product Code. 10730775

  • £92.00 / 250g


Estimated Shipment
10-07-2018
Add to basket


Specifications

Chemical Identifiers

1,2,4,5-Tetrachlorobenzene
97.5
≥97.5%
C6H2Cl4
215.89
95-94-3
100.0
98%
C6H2Cl4
MFCD00000549

Specifications

1,2,4,5-Tetrachlorobenzene
97.5
Authentic
≥97.5%
C6H2Cl4
215.89
05, 205
Light pink to gray
250g
95-94-3
100.0
Crystalline Solid
98%
C6H2Cl4
MFCD00000549
Glass bottle
139° to 142°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,2,4,5-Tetrachlorobenzene

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS