missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrachlorobenzene, 98%, Thermo Scientific Chemicals

Catalog Number 10521601
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
95-94-3
C6H2Cl4
215.89
MFCD00000549
JHBKHLUZVFWLAG-UHFFFAOYSA-N
s-tetrachlorobenzene, benzene, 1,2,4,5-tetrachloro, benzene tetrachloride, rcra waste number u207, 1,2,4,5-tetrachlorbenzol, unii-5n27529kgh, ccris 766, benzene, tetrachloro, rcra waste no. u207, 1,2,4,5-tetrachlorbenzol russian
7270
CHEBI:36697
1,2,4,5-tetrachlorobenzene
C1=C(C(=CC(=C1Cl)Cl)Cl)Cl
This item is not returnable. View return policy
95-94-3
C6H2Cl4
215.89
MFCD00000549
JHBKHLUZVFWLAG-UHFFFAOYSA-N
s-tetrachlorobenzene, benzene, 1,2,4,5-tetrachloro, benzene tetrachloride, rcra waste number u207, 1,2,4,5-tetrachlorbenzol, unii-5n27529kgh, ccris 766, benzene, tetrachloro, rcra waste no. u207, 1,2,4,5-tetrachlorbenzol russian
7270
CHEBI:36697
1,2,4,5-tetrachlorobenzene
C1=C(C(=CC(=C1Cl)Cl)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 139°C to 142°C
CAS Min % 97.5
Color Gray to Pink
CAS Max % 100.0
Boiling Point 240°C to 246°C
Quantity 1 kg
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Solid
Assay Percent Range 98%
Linear Formula C6H2Cl4
Beilstein 05, 205
Formula Weight 215.89
Percent Purity 98%
Chemical Name or Material 1,2,4,5-Tetrachlorobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1,2,4,5-Tetrachlorobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'