missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrabromobenzene, 97%, Thermo Scientific Chemicals

Catalog Number p-195465
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
636-28-2
C6H2Br4
393.70
MFCD00000063
QCKHVNQHBOGZER-UHFFFAOYSA-N
12486
1,2,4,5-tetrabromobenzene
BrC1=CC(Br)=C(Br)C=C1Br
636-28-2
C6H2Br4
393.70
MFCD00000063
QCKHVNQHBOGZER-UHFFFAOYSA-N
12486
1,2,4,5-tetrabromobenzene
BrC1=CC(Br)=C(Br)C=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 177.0°C to 182.0°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H2Br4
Quantity 25 g
Synonym benzene, 1,2,4,5-tetrabromo, 2,3,5,6-tetrabromobenzene, unii-m25dk66lcz, m25dk66lcz, pubchem4448, 1,4,5-tetrabromobenzene, 2,5,6-tetrabromobenzene, acmc-1b6z1, benzene,2,4,5-tetrabromo, benzene,1,2,4,5-tetrabromo
Solubility Information Solubility in water: insoluble.
Formula Weight 393.7
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Needles
Chemical Name or Material 1,2,4,5-Tetrabromobenzene, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1,2,4,5-Tetrabromobenzene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H413-May cause long lasting harmful effects to aquatic life.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'