missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4,5-Benzenetetracarboxylic acid, 96%, ACROS Organics™
£15.07 - £70.42
Chemical Identifiers
CAS | 89-05-4 |
---|---|
Molecular Formula | C10H6O8 |
Molecular Weight (g/mol) | 254.15 |
MDL Number | MFCD00002471 |
InChI Key | CYIDZMCFTVVTJO-UHFFFAOYSA-N |
Synonym | pyromellitic acid, 1,2,4,5-benzenetetracarboxylic acid, usaf xr-20, 1,2,4,5-tetracarboxybenzene, unii-d9uxg2u6zu, 1,2,4,5-benzene-tetracarboxylic acid, d9uxg2u6zu, pubchem21809, acmc-20ap78, 1,4,5-tetracarboxybenzene |
PubChem CID | 6961 |
ChEBI | CHEBI:45165 |
IUPAC Name | benzene-1,2,4,5-tetracarboxylic acid |
SMILES | C1=C(C(=CC(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10213990
|
Acros Organics
105310050 |
5g | Glass bottle |
£15.07
5g |
In stock, ready to ship: 6 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
10040570
|
Acros Organics
105311000 |
100g | Plastic bottle |
£21.80
100g |
In stock, ready to ship: 24 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
10132370
|
Acros Organics
105315000 |
500g | Plastic bottle |
£70.42
500g |
In stock, ready to ship: 1 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
Chemical Identifiers
89-05-4 | |
254.15 | |
CYIDZMCFTVVTJO-UHFFFAOYSA-N | |
6961 | |
benzene-1,2,4,5-tetracarboxylic acid |
C10H6O8 | |
MFCD00002471 | |
pyromellitic acid, 1,2,4,5-benzenetetracarboxylic acid, usaf xr-20, 1,2,4,5-tetracarboxybenzene, unii-d9uxg2u6zu, 1,2,4,5-benzene-tetracarboxylic acid, d9uxg2u6zu, pubchem21809, acmc-20ap78, 1,4,5-tetracarboxybenzene | |
CHEBI:45165 | |
C1=C(C(=CC(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O |
Specifications
1,2,4,5-Benzenetetracarboxylic acid | |
Glass bottle | |
281.0°C to 284.5°C | |
96% | |
95.0 | |
96% | |
C6H2(CO2H)4 | |
09, 997 | |
pyromellitic acid, 1,2,4,5-benzenetetracarboxylic acid, usaf xr-20, 1,2,4,5-tetracarboxybenzene, unii-d9uxg2u6zu, 1,2,4,5-benzene-tetracarboxylic acid, d9uxg2u6zu, pubchem21809, acmc-20ap78, 1,4,5-tetracarboxybenzene | |
CYIDZMCFTVVTJO-UHFFFAOYSA-N | |
benzene-1,2,4,5-tetracarboxylic acid | |
6961 | |
254.15 | |
96% |
Authentic | |
White to Yellow | |
5g | |
89-05-4 | |
100.0 | |
C10H6O8 | |
MFCD00002471 | |
15, 8116 | |
Solubility in water: 1.5g/100mL water (20°C). Other solubilities: 30g/100mL water (100°C),freely soluble in alcohol | |
C1=C(C(=CC(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O | |
254.15 | |
CHEBI:45165 | |
Crystalline Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1,2,4,5-Benzenetetracarboxylic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.