missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trimethoxybenzene, 98%, Thermo Scientific Chemicals

Catalog Number p-206233
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
634-36-6
C9H12O3
168.19
MFCD00008358
CRUILBNAQILVHZ-UHFFFAOYSA-N
methylsyringol, pyrogallol trimethyl ether, trimethoxybenzene, benzene, 1,2,3-trimethoxy, tri-o-methylpyrogallol, benzene, trimethoxy, unii-mre1o894fg, 1,2,3-trimethoxy benzene, mre1o894fg, 3,4,5-trimethoxybenzene
12462
CHEBI:86529
1,2,3-trimethoxybenzene
COC1=C(C(=CC=C1)OC)OC
This item is not returnable. View return policy
634-36-6
C9H12O3
168.19
MFCD00008358
CRUILBNAQILVHZ-UHFFFAOYSA-N
methylsyringol, pyrogallol trimethyl ether, trimethoxybenzene, benzene, 1,2,3-trimethoxy, tri-o-methylpyrogallol, benzene, trimethoxy, unii-mre1o894fg, 1,2,3-trimethoxy benzene, mre1o894fg, 3,4,5-trimethoxybenzene
12462
CHEBI:86529
1,2,3-trimethoxybenzene
COC1=C(C(=CC=C1)OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 42.0°C to 45.0°C
Color Beige to White
Density 1.1120g/mL
Boiling Point 241.0°C
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H3(OCH3)3
Quantity 25 g
Beilstein 06, 1081
Specific Gravity 1.112
Solubility Information Solubility in water: insoluble
Formula Weight 168.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 2, 3-Trimethoxybenzene, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'