missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Thiadiazole-4-carbaldehyde, Thermo Scientific™

Catalog Number p-4510503
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
27643-15-8
C3H2N2OS
114.122
JNEBZFFTOLBIKJ-UHFFFAOYSA-N
1,2,3-thiadiazole-4-carbaldehyde, 1,2,3-thiadiazole-4-carboxaldehyde, 4-thiadiazolecarboxaldehyde, acmc-1cb1x, 1,2,3-thiadiazol-4-carboxaldehyde
2737271
thiadiazole-4-carbaldehyde
C1=C(N=NS1)C=O
This item is not returnable. View return policy
27643-15-8
C3H2N2OS
114.122
JNEBZFFTOLBIKJ-UHFFFAOYSA-N
1,2,3-thiadiazole-4-carbaldehyde, 1,2,3-thiadiazole-4-carboxaldehyde, 4-thiadiazolecarboxaldehyde, acmc-1cb1x, 1,2,3-thiadiazol-4-carboxaldehyde
2737271
thiadiazole-4-carbaldehyde
C1=C(N=NS1)C=O
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 114.13
Percent Purity 97%
Chemical Name or Material 1, 2, 3-Thiadiazole-4-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'