missing translation for 'onlineSavingsMsg'
Learn More

1-(2,3-Dimethylphenyl)piperazine, 97%, ACROS Organics™

Brand:  Acros Organics 136930250

£30.48 valid until 30/06/2017
Log in to get the promotional price



UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 1013-22-5 Weight : 0.07500kg

Quantity 25g

 View more versions of this product

Product Code. 10316500

  • £76.20 / 25g


Estimated Shipment
21-06-2017
Add to basket


Specifications Description & Specifications

Chemical Identifiers

1-(2, 3-Dimethylphenyl)piperazine
98.5
98.5 to 101.5% Total base
C12H18N2
1013-22-5
100.0
99%
190.29

Specifications

190.29
25g
98.5
1013-22-5
99%
1-(2,3-Dimethylphenyl)piperazine
100.0
1-(2, 3-Dimethylphenyl)piperazine
Viscous Liquid
Clear light yellow
98.5 to 101.5% Total base
Authentic
1.5566 to 1.5606 (20°C, 589nm)
C12H18N2
1.0309g/mL
Documentation

Technical Documents

Special Offers

Special Offers