missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,3-Dimethylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64501.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15458956

  • £298.92 / 5g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,3-Dimethylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
324.3
MFCD07096485

Specifications

1-(2,3-Dimethylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
324.3
5g
97%
MFCD07096485
Videos
SDS