missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,3-Dimethylphenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62916.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15419368

  • £266.06 / 5g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,3-Dimethylphenyl)-3-(4-fluorophenyl)urea
258.29
MFCD06121910

Specifications

1-(2,3-Dimethylphenyl)-3-(4-fluorophenyl)urea
Solid
258.29
5g
97%
MFCD06121910
Videos
SDS