missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,3-Dichlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64883.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15499226

  • £610.56 / 5g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,3-Dichlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
365.13
MFCD04281090

Specifications

1-(2,3-Dichlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
365.13
5g
97%
MFCD04281090
Videos
SDS