Learn More
1,2,3-Benzothiadiazole-5-carbonyl chloride, 95%, Maybridge™
Brand: Maybridge CC09402DE
Code : 31
Additional Details : CAS Number : 321309-32-4 Weight : 0.05500kg Transport : UN number : 3261 Chem class : 8 Pack group : III
Quantity | 5g |
---|
Chemical Identifiers
321309-32-4 | |
198.624 | |
1,2,3-benzothiadiazole-5-carbonylchloride, benzo d 1,2,3-thiadiazole-5-carbonyl chloride, benzo d 1,2,3 thiadiazole-5-carbonyl chloride | |
1,2,3-benzothiadiazole-5-carbonyl chloride |
C7H3ClN2OS | |
VOSTUOLSYXVSND-UHFFFAOYSA-N | |
2735457 | |
C1=CC2=C(C=C1C(=O)Cl)N=NS2 |
Specifications
1, 2, 3-Benzothiadiazole-5-carbonyl chloride | |
5g | |
95% | |
1,2,3-benzothiadiazole-5-carbonylchloride, benzo d 1,2,3-thiadiazole-5-carbonyl chloride, benzo d 1,2,3 thiadiazole-5-carbonyl chloride | |
C1=CC2=C(C=C1C(=O)Cl)N=NS2 | |
198.624 | |
198.63 |
Amber Glass Bottle | |
321309-32-4 | |
C7H3ClN2OS | |
VOSTUOLSYXVSND-UHFFFAOYSA-N | |
1,2,3-benzothiadiazole-5-carbonyl chloride | |
2735457 | |
95% |