missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydronaphthalene, Reagent Grade, ≥97%, Honeywell™

Catalog Number p-7120009
Click to view available options
Quantity:
20 L
Packaging:
Stainless Steel Can
This item is not returnable. View return policy
119-64-2
C10H12
132.21
MFCD00001733
CXWXQJXEFPUFDZ-UHFFFAOYSA-N
tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene
8404
CHEBI:35008
1,2,3,4-tetrahydronaphthalene
C1CCC2=CC=CC=C2C1
This item is not returnable. View return policy
119-64-2
C10H12
132.21
MFCD00001733
CXWXQJXEFPUFDZ-UHFFFAOYSA-N
tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene
8404
CHEBI:35008
1,2,3,4-tetrahydronaphthalene
C1CCC2=CC=CC=C2C1
Melting Point -35°C
Color Colorless
Density 0.973g/mL at 25°C
Boiling Point 207°C
Flash Point 71°C (159.8°F)
Infrared Spectrum Conforms to structure
Packaging Stainless Steel Can
Refractive Index n20/D 1.541
Quantity 20 L
UN Number UN3082
Beilstein 1446407
Formula Weight 132.2g/mol
Percent Purity ≥97%
Chemical Name or Material 1,2,3,4-Tetrahydronaphthalene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'