missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydrobenzazepine-5-one, 97%, Thermo Scientific™

Catalog Number p-4228904
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1127-74-8
MFCD03426404
NKRKBYFBKLDCFB-UHFFFAOYSA-N
1,2,3,4-tetrahydro-benzo b azepin-5-one, 1,2,3,4-tetrahydrobenzo b azepin-5-one, 1,2,3,4-tetrahydrobenzoazepine-5-one, 3,4-dihydro-1h-benzo b azepin-5 2h-one, 1,2,3,4-tetrahydro-5h-1-benzazepin-5-one, 2,3,4,5-tetrahydro-1h-1-benzazepin-5-one, 5h-1-benzazepin-5-one, 1,2,3,4-tetrahydro, 5-oxo-2,3,4,5-tetrahydro-1h-benzazepine, 1,2,3,4-tetrahydrobenzazepine-5-one, zlchem 1002
294788
1,2,3,4-tetrahydro-1-benzazepin-5-one
C1CC(=O)C2=CC=CC=C2NC1
This item is not returnable. View return policy
1127-74-8
MFCD03426404
NKRKBYFBKLDCFB-UHFFFAOYSA-N
1,2,3,4-tetrahydro-benzo b azepin-5-one, 1,2,3,4-tetrahydrobenzo b azepin-5-one, 1,2,3,4-tetrahydrobenzoazepine-5-one, 3,4-dihydro-1h-benzo b azepin-5 2h-one, 1,2,3,4-tetrahydro-5h-1-benzazepin-5-one, 2,3,4,5-tetrahydro-1h-1-benzazepin-5-one, 5h-1-benzazepin-5-one, 1,2,3,4-tetrahydro, 5-oxo-2,3,4,5-tetrahydro-1h-benzazepine, 1,2,3,4-tetrahydrobenzazepine-5-one, zlchem 1002
294788
1,2,3,4-tetrahydro-1-benzazepin-5-one
C1CC(=O)C2=CC=CC=C2NC1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 70.0°C to 71.0°C
Color Green to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C10H11NO
Quantity 1 g
Molecular Weight (g/mol) 161.21
Formula Weight 161.21
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,2,3,4-Tetrahydrobenzazepine-5-one
Show More Show Less
compliance-icons
Product Identifier
  • 1,2,3,4-Tetrahydrobenzazepine-5-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'