missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydro-1-naphthylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-3830559
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2217-40-5
C10H14ClN
183.68
MFCD00001740
DETWFIUAXSWCIK-UHFFFAOYNA-N
1,2,3,4-tetrahydro-1-naphthylamine, 1-aminotetralin, aminotetralin, 1-amino-1,2,3,4-tetrahydro-naphthalene, aminotetralin czech, tetrahydro-beta-naphthylamine, 1-amino-1,2,3,4-tetrahydronaphthalene, 1-naphthalenamine, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydronaphthylamine, beta-1,2,3,4-tetrahydronaphthylamine
18066
1,2,3,4-tetrahydronaphthalen-1-amine
[Cl-].[NH3+]C1CCCC2=CC=CC=C12
This item is not returnable. View return policy
2217-40-5
C10H14ClN
183.68
MFCD00001740
DETWFIUAXSWCIK-UHFFFAOYNA-N
1,2,3,4-tetrahydro-1-naphthylamine, 1-aminotetralin, aminotetralin, 1-amino-1,2,3,4-tetrahydro-naphthalene, aminotetralin czech, tetrahydro-beta-naphthylamine, 1-amino-1,2,3,4-tetrahydronaphthalene, 1-naphthalenamine, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydronaphthylamine, beta-1,2,3,4-tetrahydronaphthylamine
18066
1,2,3,4-tetrahydronaphthalen-1-amine
[Cl-].[NH3+]C1CCCC2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0200g/mL
Boiling Point 246.0°C to 247.0°C (714.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5625 to 1.5645
Quantity 5 g
Beilstein 12,12
Specific Gravity 1.02
Formula Weight 147.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1,2,3,4-Tetrahydro-1-naphthylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'