missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2:3,4-Di-O-isopropylidene-D-galactopyranose, 97%

Catalog Number p-113613
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4064-06-6
C12H20O6
260.28
MFCD00063225
POORJMIIHXHXAV-UHFFFAOYNA-N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4064-06-6
C12H20O6
260.28
MFCD00063225
POORJMIIHXHXAV-UHFFFAOYNA-N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 120.0°C to 122.0°C
Color Yellow
Density 1.1400g/mL
Boiling Point 117.0°C (0.1 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4668 to 1.4688
Quantity 5 g
Specific Gravity 1.14
Specific Rotation Condition − 57.50 (20.00°C c=3,CHCl3)
Specific Rotation -57.5
Synonym 1,2:3,4-di-o-isopropylidene-d-galactose, 1,2:3,4-di-o-isopropylidene-, a-d-galactopyranose, 1,2:3,4-bis-o-1-methylethylidene-, 1s,6r,9s-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo 7.3.0.0 2 ,? dodecan-8-yl methanol
SMILES CC1(OC2C(OC3C(C2O1)OC(O3)(C)C)CO)C
IUPAC Name [(3aR,5aS,8aS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydro-3aH-di[1,3]dioxolo[4,5-a
PubChem CID 70793572
Formula Weight 260.28
Percent Purity 97%
Physical Form Viscous Paste
Chemical Name or Material 1, 2:3, 4-Di-O-isopropylidene-D-galactopyranose, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'