missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,5-Pentamethylcyclopentadiene, 95%, Thermo Scientific™

£43.90 - £414.00

Chemical Identifiers

CAS 4045-44-7
Molecular Formula C10H16
Molecular Weight (g/mol) 136.11
MDL Number MFCD00001354
InChI Key WQIQNKQYEUMPBM-UHFFFAOYSA-N
Synonym 1,2,3,4,5-pentamethylcyclopentadiene, pentamethylcyclopentadiene, cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, pentamethyl, 1,2,3,4,5-pentamethyl-1,3-cyclopentadiene, pubchem20101, acmc-209je4, wqiqnkqyeumpbm-uhfffaoysa, 1,2,3,4,5-pentamethylcyclopentadien
PubChem CID 77667
IUPAC Name 1,2,3,4,5-pentamethylcyclopenta-1,3-diene
SMILES CC1C(=C(C(=C1C)C)C)C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10484985
SDS View Documents
Thermo Scientific Acros
320830010
1mL Glass bottle
£43.90
1mL
View alternative products
Save Up to 
»
In stock, ready to ship: 1
Please to purchase this item. Need a web account? Register with us today!
10267570
SDS View Documents
Thermo Scientific Acros
320830050
5mL Glass bottle
£129.00
5mL
View alternative products
Save Up to 
»
In stock, ready to ship: 1
Please to purchase this item. Need a web account? Register with us today!
10115313
SDS View Documents
Thermo Scientific Acros
320830250
25mL Glass bottle
£414.00
25mL
View alternative products
Save Up to 
»
In stock, ready to ship: 7
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

4045-44-7
136.11
WQIQNKQYEUMPBM-UHFFFAOYSA-N
77667
CC1C(=C(C(=C1C)C)C)C
C10H16
MFCD00001354
1,2,3,4,5-pentamethylcyclopentadiene, pentamethylcyclopentadiene, cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, pentamethyl, 1,2,3,4,5-pentamethyl-1,3-cyclopentadiene, pubchem20101, acmc-209je4, wqiqnkqyeumpbm-uhfffaoysa, 1,2,3,4,5-pentamethylcyclopentadien
1,2,3,4,5-pentamethylcyclopenta-1,3-diene

Specifications

1,2,3,4,5-Pentamethylcyclopentadiene
44°C
Glass bottle
0.87
Colorless to Orange-Yellow
95%
94.0
94% min. (GC)
MFCD00001354
WQIQNKQYEUMPBM-UHFFFAOYSA-N
1,2,3,4,5-pentamethylcyclopenta-1,3-diene
77667
Liquid
0.8700g/mL
Authentic
1.4725 to 1.4755
58°C (13.0mmHg)
1mL
4045-44-7
100.0
C10H16
1,2,3,4,5-pentamethylcyclopentadiene, pentamethylcyclopentadiene, cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, pentamethyl, 1,2,3,4,5-pentamethyl-1,3-cyclopentadiene, pubchem20101, acmc-209je4, wqiqnkqyeumpbm-uhfffaoysa, 1,2,3,4,5-pentamethylcyclopentadien
CC1C(=C(C(=C1C)C)C)C
136.11
136.11
95%
SDS
Product Certifications

RUO – Research Use Only