missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,5-Pentamethylcyclopentadiene, 95%, ACROS Organics™

Brand:  Acros Organics 320830050

Code : 29

Additional Details : CAS Number : 4045-44-7 Weight : 0.05435kg Transport : UN number : 1993 Chem class : 3 Pack group : III

Quantity 5mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10267570

  • £116.90 / 5mL


Estimated Shipment
13-06-2018
Add to basket

Specifications

Chemical Identifiers

1,2,3,4,5-Pentamethylcyclopentadiene
4045-44-7
100.0
95%
136.11
95%
94.0
≥94% (GC)
C10H16
MFCD00001354

Specifications

1,2,3,4,5-Pentamethylcyclopentadiene
Clear colorless to yellow-orange, may darken on storage
95%
94.0
Authentic
1.4720 to 1.4760 (20°C, 589nm)
≥94% (GC)
C10H16
MFCD00001354
0.87g/mL
5mL
4045-44-7
100.0
Glass bottle
Liquid
95%
136.11
Videos
SDS