missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(1H-Pyrazol-5-yl)ethan-1-one hydrochloride, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Quantity | 5 g |
Formula Weight | 146.58 |
Percent Purity | 95% |
Chemical Name or Material | 1-(1H-pyrazol-5-yl)ethan-1-one hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'