missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), complex with dichloromethane (1:1), Pd 13%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II) is used for palladium-catalyzed coupling reactions. It serves as a catalyst and also employed in carbonylation reactions. Further, it is used in cross-coupling reaction and Suzuki reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 275°C to 280°C |
Quantity | 1 g |
Synonym | Dichloro[1, 1′-bis(diphenylphosphino)ferrocene]palladium(II) dichloromethane complex |
Solubility Information | Insoluble in water. |
Formula Weight | 816.64 |
Physical Form | Powder |
Chemical Name or Material | [1, 1′-Bis(diphenylphosphino)ferrocene]palladium(II) chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'