missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,12-Dodecanediol, 98%, ACROS Organics™
£26.92 - £122.00
Chemical Identifiers
CAS | 5675-51-4 |
---|---|
Molecular Formula | C12H26O2 |
Molecular Weight (g/mol) | 202.34 |
InChI Key | GHLKSLMMWAKNBM-UHFFFAOYSA-N |
Synonym | 1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h |
PubChem CID | 79758 |
IUPAC Name | dodecane-1,12-diol |
SMILES | C(CCCCCCO)CCCCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
13488318
|
Acros Organics
117610250 |
25g | Glass bottle |
£26.92
25g |
In stock, ready to ship: 3 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
13428328
|
Acros Organics
117611000 |
100g | Glass bottle |
£36.37
100g |
In stock, ready to ship: 5 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
13478318
|
Acros Organics
117610010 |
1kg | Glass bottle |
£122.00
1kg |
In stock, ready to ship: 3 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
Chemical Identifiers
5675-51-4 | |
202.34 | |
1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h | |
dodecane-1,12-diol |
C12H26O2 | |
GHLKSLMMWAKNBM-UHFFFAOYSA-N | |
79758 | |
C(CCCCCCO)CCCCCO |
Specifications
1, 12-Dodecanediol | |
Authentic | |
16 mPa.s (85°C) | |
Brown to Orange | |
25g | |
5675-51-4 | |
100.0 | |
C12H26O2 | |
01,II,562 | |
Solubility in water: <0.1% in water. | |
C(CCCCCCO)CCCCCO | |
202.34 | |
202.34 | |
98% |
176°C | |
Glass bottle | |
324.0°C | |
79.0°C to 81.0°C | |
99% | |
98.5 | |
97.5% min. (GC) | |
HO(CH2)12OH | |
1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h | |
GHLKSLMMWAKNBM-UHFFFAOYSA-N | |
dodecane-1,12-diol | |
79758 | |
Powder |