missing translation for 'onlineSavingsMsg'
Learn More

1,12-Dodecanediol, 98%, Thermo Scientific Chemicals

Catalog Number 13488318
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
5675-51-4
C12H26O2
202.34
MFCD00004755
GHLKSLMMWAKNBM-UHFFFAOYSA-N
1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h
79758
dodecane-1,12-diol
OCCCCCCCCCCCCO
This item is not returnable. View return policy
5675-51-4
C12H26O2
202.34
MFCD00004755
GHLKSLMMWAKNBM-UHFFFAOYSA-N
1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h
79758
dodecane-1,12-diol
OCCCCCCCCCCCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 79.0°C to 81.0°C
Color Brown to Orange
Boiling Point 324.0°C
Flash Point 176°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HO(CH2)12OH
Quantity 25 g
Beilstein 01,II,562
Solubility Information Solubility in water: <0.1% in water.
Viscosity 16 mPa.s (85°C)
Formula Weight 202.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 12-Dodecanediol, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'