missing translation for 'onlineSavingsMsg'
Learn More

1,12-Dodecanediol, 98%, ACROS Organics™

Brand:  Acros Organics 117611000

Code : NEW

Additional Details : CAS Number : 5675-51-4 Weight : 0.10000kg

Packaging Glass bottle
Quantity 100g

 View more versions of this product

Product Code. 13428328

  • £37.61 / 100g

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

5675-51-4
202.34
GHLKSLMMWAKNBM-UHFFFAOYSA-N
79758
OCCCCCCCCCCCCO
C12H26O2
MFCD00004755
1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h
dodecane-1,12-diol

Specifications

1, 12-Dodecanediol
Authentic
16 mPa.s (85°C)
Brown to Orange
100g
5675-51-4
100.0
C12H26O2
MFCD00004755
1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h
GHLKSLMMWAKNBM-UHFFFAOYSA-N
dodecane-1,12-diol
79758
Powder
176°C
Glass bottle
324.0°C
79.0°C to 81.0°C
99%
98.5
97.5% min. (GC)
HO(CH2)12OH
01,II,562
Solubility in water: <0.1% in water.
OCCCCCCCCCCCCO
202.34
202.34
98%
Videos
SDS Documents
Certificates