missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Thiocarbonyldiimidazole, 90%, Tech., Thermo Scientific Chemicals

Catalog Number 10686372
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6160-65-2
C7H6N4S
178.21
MFCD00005289
RAFNCPHFRHZCPS-UHFFFAOYSA-N
1,1'-thiocarbonyldiimidazole, thiocarbonyldiimidazole, n,n'-thiocarbonyldiimidazole, di 1h-imidazol-1-yl methanethione, 1,1-thiocarbonyldiimidazole, di imidazol-1-yl methanethione, n,n'-thiocarbonyldiimidazole tcdi, 1h-imidazole, 1,1'-carbonothioylbis, n,n-thiocarbonyldiimidazole, 1,1'-carbonothioylbis-1h-imidazole
80264
di(imidazol-1-yl)methanethione
S=C(N1C=CN=C1)N1C=CN=C1
This item is not returnable. View return policy
6160-65-2
C7H6N4S
178.21
MFCD00005289
RAFNCPHFRHZCPS-UHFFFAOYSA-N
1,1'-thiocarbonyldiimidazole, thiocarbonyldiimidazole, n,n'-thiocarbonyldiimidazole, di 1h-imidazol-1-yl methanethione, 1,1-thiocarbonyldiimidazole, di imidazol-1-yl methanethione, n,n'-thiocarbonyldiimidazole tcdi, 1h-imidazole, 1,1'-carbonothioylbis, n,n-thiocarbonyldiimidazole, 1,1'-carbonothioylbis-1h-imidazole
80264
di(imidazol-1-yl)methanethione
S=C(N1C=CN=C1)N1C=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 88.0
CAS Max % 100.0
Melting Point 96.0°C to 103.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Quantity 10 g
Fieser 01,1151; 02,411; 05,661; 06,583; 13,318
Solubility Information Solubility in water: decomposes.
Formula Weight 178.21
Percent Purity 90%
Grade Technical
Physical Form Powder
Chemical Name or Material 1, 1'-Thiocarbonyldiimidazole
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,1'-Thiocarbonylbis(imidazole)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'