missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Naphthyl)piperazine hydrochloride, Thermo Scientific Chemicals

Catalog Number p-7042313
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
104113-71-5
C14H17ClN2
248.754
MFCD00055161
ZYVYPNZFOCZLEM-UHFFFAOYSA-N
1-1-naphthyl piperazine hydrochloride, 1-naphthalen-1-yl piperazine hydrochloride, naphthylpiperazine hydrochloride, 1-naphthalen-1-ylpiperazine hydrochloride, 1-naphthalen-1-yl-piperazine hydrochloride, piperazine,1-1-naphthalenyl, zlchem 1062, 1-1-naphthyl piperazine hcl, 1-naphthalen-1-yl-piperazine hcl salt, n-1-naphthyl piperazine hydrochloride
2737391
1-naphthalen-1-ylpiperazine;hydrochloride
C1CN(CCN1)C2=CC=CC3=CC=CC=C32.Cl
This item is not returnable. View return policy
104113-71-5
C14H17ClN2
248.754
MFCD00055161
ZYVYPNZFOCZLEM-UHFFFAOYSA-N
1-1-naphthyl piperazine hydrochloride, 1-naphthalen-1-yl piperazine hydrochloride, naphthylpiperazine hydrochloride, 1-naphthalen-1-ylpiperazine hydrochloride, 1-naphthalen-1-yl-piperazine hydrochloride, piperazine,1-1-naphthalenyl, zlchem 1062, 1-1-naphthyl piperazine hcl, 1-naphthalen-1-yl-piperazine hcl salt, n-1-naphthyl piperazine hydrochloride
2737391
1-naphthalen-1-ylpiperazine;hydrochloride
C1CN(CCN1)C2=CC=CC3=CC=CC=C32.Cl

5-HT1 serotonin receptor agonist. 5-HT2 serotonin receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • 5-HT1 serotonin receptor agonist
  • 5-HT2 serotonin receptor antagonist
TRUSTED_SUSTAINABILITY
Color Purple
Quantity 1 g
Formula Weight 248.75
Physical Form Solid
Chemical Name or Material 1-(1-Naphthyl)piperazine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'