missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Naphthyl)-2-(trimethylsilyl)acetylene, 97%, Thermo Scientific Chemicals

Catalog Number p-7052813
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
104784-51-2
C15H16Si
224.378
MFCD04039886
WATBCTJRCLNXNF-UHFFFAOYSA-N
1-1-naphthyl-2-trimethylsilyl acetylene, trimethyl 2-naphthalen-1-ylethynyl silane, trimethyl 2-naphthalen-1-yl ethynyl silane, 1-naphthylethynyl trimethylsilane, 1-trimethylsilylethynyl naphthalene, 1-trimethylsilyl ethynyl naphthalene, trimethyl naphthalen-1-ylethynyl silane, trimethyl naphthalen-1-yl ethynyl silane, trimethylnaphthalen-1-ylethynylsilane
4438232
trimethyl(2-naphthalen-1-ylethynyl)silane
C[Si](C)(C)C#CC1=CC=CC2=CC=CC=C21
This item is not returnable. View return policy
104784-51-2
C15H16Si
224.378
MFCD04039886
WATBCTJRCLNXNF-UHFFFAOYSA-N
1-1-naphthyl-2-trimethylsilyl acetylene, trimethyl 2-naphthalen-1-ylethynyl silane, trimethyl 2-naphthalen-1-yl ethynyl silane, 1-naphthylethynyl trimethylsilane, 1-trimethylsilylethynyl naphthalene, 1-trimethylsilyl ethynyl naphthalene, trimethyl naphthalen-1-ylethynyl silane, trimethyl naphthalen-1-yl ethynyl silane, trimethylnaphthalen-1-ylethynylsilane
4438232
trimethyl(2-naphthalen-1-ylethynyl)silane
C[Si](C)(C)C#CC1=CC=CC2=CC=CC=C21

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 41°C to 43°C
Density 0.968 g/mL
Boiling Point 292°C to 293°C
Flash Point >110°C (230°F)
Refractive Index 1.595
Quantity 1 g
Formula Weight 224.38
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(1-Naphthyl)-2-(trimethylsilyl)acetylene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'