missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(1-Naphthyl)-2-(trimethylsilyl)acetylene, 97%

Brand:  Alfa Aesar™ H55953.06

Code : NEW

Additional Details : CAS Number : 104784-51-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15442358

  • £73.50 / 5g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

1-(1-Naphthyl)-2-(trimethylsilyl)acetylene
224.38
Trimethyl[(1-naphthyl)ethynyl]silane
104784-51-2
MFCD04039886

Specifications

1-(1-Naphthyl)-2-(trimethylsilyl)acetylene
1.5950
97%
224.38
MFCD04039886
5g
104784-51-2
Solid
0.968g/mL
41° to 43°C
292° to 293°C
Trimethyl[(1-naphthyl)ethynyl]silane
Videos
SDS