missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylurea, 98+%, Thermo Scientific Chemicals

Catalog Number p-3527043
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
598-94-7
C3H8N2O
88.11
MFCD00007959
YBBLOADPFWKNGS-UHFFFAOYSA-N
n,n-dimethylurea, urea, n,n-dimethyl, asym-dimethylurea, urea, dimethyl, urea, 1,1-dimethyl, n,n-dimethylharnstoff german, n,n-dimethylharnstoff, dimethyl urea, 1.1-dimethylurea, 1,1-dimethyl-urea
11737
1,1-dimethylurea
CN(C)C(=O)N
This item is not returnable. View return policy
598-94-7
C3H8N2O
88.11
MFCD00007959
YBBLOADPFWKNGS-UHFFFAOYSA-N
n,n-dimethylurea, urea, n,n-dimethyl, asym-dimethylurea, urea, dimethyl, urea, 1,1-dimethyl, n,n-dimethylharnstoff german, n,n-dimethylharnstoff, dimethyl urea, 1.1-dimethylurea, 1,1-dimethyl-urea
11737
1,1-dimethylurea
CN(C)C(=O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 179°C to 183°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (CH3)2NC(O)NH2
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 88.11
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 1, 1-Dimethylurea
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'